Search
Search

SEC14L1| Gene Symbol: SEC14L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039573.2 | NP_001034662.2 | 15 | 1981 | ||
| NM_001143998.1 | NP_001137470.1 | 15 | 1981 | |||
| NM_001143999.1 | NP_001137471.1 | 15 | 1977 | |||
| NM_001144001.1 | NP_001137473.1 | 13 | 1769 | |||
| NM_001204408.1 | NP_001191337.1 | 17 | 2326 | |||
| NM_001204410.1 | NP_001191339.1 | 16 | 2019 | |||
| NM_003003.3 | NP_002994.3 | 15 | 1928 | |||
| GenBank mRNA | AB208811.1 | - | 15 | 1960 | ||
| AK094232.1 | - | 1 | 2139 | |||
| AK130317.1 | - | 2 | 681 | |||
| AK290913.1 | - | 15 | 1874 | |||
| AK293206.1 | - | 13 | 1769 | |||
| AK293867.1 | - | 3 | 221 | |||
| BC136523.1 | - | 15 | 1816 | 9020513 | ||
| BC136525.1 | - | 15 | 1816 | 9020515 | ||
| BC142979.1 | - | 15 | 1872 | 8860414 | ||
| BX537448.1 | - | 15 | 1882 | |||
| D67029.1 | - | 15 | 1964 | |||
| FM995495.1 | - | 17 | 2220 | |||
| FM995496.1 | - | 16 | 2017 | |||
| U79284.1 | - | 4 | 397 | |||