Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ajuba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053503.1 | NP_445955.1 | 2 | 1371 | ||
GenBank mRNA | AJ306292.1 | - | 2 | 521 | ||
BC085802.1 | - | 2 | 1371 | 7110970 |