Search
Search

Alg3| Gene Symbol: Alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142363.1 | NP_001135835.1 | 3 | 403 | ||
| XM_006248558.2 | XP_006248620.1 | 3 | 421 | |||
| GenBank mRNA | BC088475.1 | - | 2 | 286 | 7129249 | |
| CO401508.1 | - | 3 | 392 | |||
| FM075565.1 | - | 2 | 303 | |||