Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1G SIRNA Assays ›
Gene Symbol:
CACNA1G
Gene Name:
calcium voltage-gated channel subunit alpha1 G
Gene Aliases:
Ca(V)T.1, Cav3.1, NBR13, SCA42
Chromosome Location:
Chr.17: 50560929 - 50629625 on Build GRCh38
UniGene ID:
Hs.591169
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17050
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256324.1
NP_001243253.1
7
1496
NM_001256325.1
NP_001243254.1
7
1496
NM_001256326.1
NP_001243255.1
7
1496
NM_001256327.1
NP_001243256.1
7
1496
NM_001256328.1
NP_001243257.1
7
1496
NM_001256329.1
NP_001243258.1
7
1496
NM_001256330.1
NP_001243259.1
7
1496
NM_001256331.1
NP_001243260.1
7
1496
NM_001256332.1
NP_001243261.1
7
1496
NM_001256333.1
NP_001243262.1
7
1496
NM_001256334.1
NP_001243263.1
7
1496
NM_001256359.1
NP_001243288.1
7
1496
NM_001256360.1
NP_001243289.1
7
1496
NM_001256361.1
NP_001243290.1
7
1496
NM_018896.4
NP_061496.2
7
1496
NM_198376.2
NP_938190.1
7
1496
NM_198377.2
NP_938191.2
7
1496
NM_198378.2
NP_938192.1
7
1496
NM_198379.2
NP_938193.1
7
1496
NM_198380.2
NP_938194.1
7
1496
NM_198382.2
NP_938196.1
7
1496
NM_198383.2
NP_938197.1
7
1496
NM_198384.2
NP_938198.1
7
1496
NM_198385.2
NP_938199.1
7
1496
NM_198386.2
NP_938200.1
7
1496
NM_198387.2
NP_938201.1
7
1496
NM_198388.2
NP_938202.1
7
1496
NM_198396.2
NP_938406.1
7
1496
NR_046054.1
7
1496
NR_046055.1
7
1496
NR_046056.1
7
1496
NR_046057.1
7
1496
NR_046058.1
7
1496
XM_006722160.3
XP_006722223.1
7
1635
XM_006722161.3
XP_006722224.1
7
1635
GenBank mRNA
AB012043.1
-
7
1132
AF124351.1
-
7
1476
AF126965.1
-
7
1279
AF126966.1
-
7
1279
AF134985.1
-
7
1104
AF134986.1
-
7
1104
AF190860.1
-
7
1320
AF227744.1
-
7
1104
AF227745.1
-
7
1104
AF227746.1
-
7
1104
AF227747.1
-
7
1104
AF227748.1
-
7
1104
AF227749.1
-
7
1104
AF227750.1
-
7
1104
AF227751.1
-
7
1104
BC110995.1
-
7
1496
5502027
DQ494449.1
-
7
1104
DQ494450.1
-
7
1104
DQ494451.1
-
7
1104
DQ494452.1
-
7
1104
DQ494453.1
-
7
1104
DQ494454.1
-
7
1104
DQ494455.1
-
7
1104
DQ494456.1
-
7
1104
DQ494457.1
-
7
1104
DQ494458.1
-
7
1104
DQ494459.1
-
7
1104
DQ494460.1
-
7
1104
DQ494461.1
-
7
1104
DQ494462.1
-
7
1104
DQ494463.1
-
7
1104
DQ494464.1
-
7
1104
DQ494465.1
-
7
1104
DQ494466.1
-
7
1104
DQ494467.1
-
7
1104
DQ494468.1
-
7
1104
DQ494469.1
-
7
1104
DQ494470.1
-
7
1104
DQ494471.1
-
7
1104
DQ494472.1
-
7
1104
DQ494473.1
-
7
1104
DQ494474.1
-
7
1104
DQ494475.1
-
7
1104
DQ494476.1
-
7
1104
DQ494477.1
-
7
1104
DQ494478.1
-
7
1104
DQ494479.1
-
7
1104
KJ534993.1
-
7
1104
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
response to nickel cation
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of calcium ion-dependent exocytosis
regulation of atrial cardiac muscle cell membrane depolarization
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
SA node cell action potential
AV node cell action potential
SA node cell to atrial cardiac muscle cell signalling
AV node cell to bundle of His cell signaling
membrane depolarization during AV node cell action potential
membrane depolarization during SA node cell action potential
regulation of heart rate by cardiac conduction
low voltage-gated calcium channel activity
voltage-gated calcium channel activity involved in AV node cell action potential
voltage-gated calcium channel activity involved SA node cell action potential
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0