Search
Search

| Gene Symbol: Olr1435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000777.1 | NP_001000777.1 | 1 | 517 | ||
| Gene Symbol: Olr1442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000776.1 | NP_001000776.1 | 1 | 517 | ||