Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SRSF11 SIRNA Assays ›
          Gene Symbol:
          SRSF11
          Gene Name:
          serine and arginine rich splicing factor 11
          Gene Aliases:
          NET2, SFRS11, dJ677H15.2, p54
          Chromosome Location:
          Chr.1: 70205682 - 70252020 on Build GRCh38
          UniGene ID:
          Hs.479693
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s17769

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SRSF11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190987.1 NP_001177916.1 3 440
          NM_004768.3 NP_004759.1 3 443
          XM_005271338.3 XP_005271395.1 3 434
          XM_005271339.3 XP_005271396.1 3 436
          XM_011542427.2 XP_011540729.1 2 596
          XM_011542428.2 XP_011540730.1 2 596
          XM_011542431.2 XP_011540733.1 3 423
          XM_017002811.1 XP_016858300.1 3 434
          XM_017002812.1 XP_016858301.1 3 434
          XM_017002813.1 XP_016858302.1 3 436
          XM_017002815.1 XP_016858304.1 2 596
          XM_017002816.1 XP_016858305.1 3 367
          XM_017002817.1 XP_016858306.1 2 596
          XM_017002818.1 XP_016858307.1 2 596
          XM_017002819.1 XP_016858308.1 2 596
          XM_017002820.1 XP_016858309.1 2 596
          XM_017002821.1 XP_016858310.1 2 596
          XM_017002824.1 XP_016858313.1 3 367
          XM_017002825.1 XP_016858314.1 2 596
          XM_017002826.1 XP_016858315.1 2 596
          XM_017002827.1 XP_016858316.1 2 596
          XM_017002828.1 XP_016858317.1 3 367
          XM_017002829.1 XP_016858318.1 3 403
          XM_017002830.1 XP_016858319.1 3 397
          XM_017002831.1 XP_016858320.1 2 596
          XM_017002832.1 XP_016858321.1 2 596
          XM_017002833.1 XP_016858322.1 2 596
          XM_017002834.1 XP_016858323.1 2 1555
          GenBank mRNA AK021742.1 - 1 1531
          AK300147.1 - 3 387
          AK301670.1 - 2 494
          AK310447.1 - 3 398
          AK310666.1 - 3 367
          AK311184.1 - 2 173
          BC002784.2 - 2 454
          BC009840.2 - 2 437
          BC017359.2 - 2 560
          BC022815.1 - 3 440
          BC032727.1 - 3 427
          BC040436.1 - 3 398 5518580
          M74002.1 - 3 402

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001035255, XM_006233540, XM_006233542, XM_008761568, XM_006233541, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001093753, NM_001093752, BC069945, AK011472, AK040467, and others.

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          termination of RNA polymerase II transcription
          mRNA processing
          RNA export from nucleus
          mRNA export from nucleus
          RNA splicing
          mRNA 3'-end processing
          nucleotide binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0