Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC4A7 SIRNA Assays ›
          Gene Symbol:
          SLC4A7
          Gene Name:
          solute carrier family 4 member 7
          Gene Aliases:
          NBC2, NBC3, NBCN1, SBC2, SLC4A6
          Chromosome Location:
          Chr.3: 27372721 - 27484420 on Build GRCh38
          UniGene ID:
          Hs.250072
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s18207

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC4A7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001258379.1 NP_001245308.1 5, 6 876
          NM_001258380.1 NP_001245309.1 5, 6 638
          NM_001321103.1 NP_001308032.1 5, 6 888
          NM_001321104.1 NP_001308033.1 5, 6 888
          NM_001321105.1 NP_001308034.1 5, 6 888
          NM_001321106.1 NP_001308035.1 5, 6 876
          NM_001321107.1 NP_001308036.1 5, 6 876
          NM_001321108.1 NP_001308037.1 5, 6 888
          NM_003615.4 NP_003606.3 5, 6 638
          NR_135541.1 5, 6 876
          NR_135542.1 5, 6 876
          NR_135543.1 5, 6 876
          NR_135544.1 5, 6 876
          XM_005265598.4 XP_005265655.1 5, 6 1142
          XM_005265600.4 XP_005265657.1 5, 6 1143
          XM_006713421.2 XP_006713484.1 5, 6 876
          XM_011534256.2 XP_011532558.1 5, 6 1149
          XM_011534258.1 XP_011532560.1 5, 6 852
          XM_011534261.2 XP_011532563.1 5, 6 1153
          XM_011534263.2 XP_011532565.1 5, 6 852
          XM_011534265.1 XP_011532567.1 5, 6 852
          XM_017007526.1 XP_016863015.1 6, 7 773
          XM_017007527.1 XP_016863016.1 5, 6 876
          GenBank mRNA AB012130.1 - 2, 3 324
          AF047033.1 - 5, 6 638
          AF053755.1 - 2, 3 316
          AF089726.1 - 5, 6 638
          AK309734.1 - 5, 6 876
          EU499349.1 - 5, 6 803
          EU934246.1 - 5, 6 803
          EU934247.1 - 5, 6 815
          EU934248.1 - 5, 6 815
          EU934249.1 - 5, 6 803
          EU934250.1 - 5, 6 815
          FJ178574.1 - 5, 6 803
          FJ178575.1 - 5, 6 803
          FJ178576.1 - 5, 6 803
          GU354307.1 - 5, 6 803
          GU354308.1 - 5, 6 815
          GU354309.1 - 5, 6 815
          GU354310.1 - 5, 6 815

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          inorganic anion transport
          bicarbonate transport
          sodium ion transmembrane transport
          regulation of intracellular pH
          auditory receptor cell development
          anion transmembrane transport
          inorganic anion exchanger activity
          anion transmembrane transporter activity
          sodium:bicarbonate symporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0