Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NR1I3 SIRNA Assays ›
Gene Symbol:
NR1I3
Gene Name:
nuclear receptor subfamily 1 group I member 3
Gene Aliases:
CAR, CAR1, MB67
Chromosome Location:
Chr.1: 161229666 - 161238623 on Build GRCh38
UniGene ID:
Hs.349642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19370
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NR1I3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077469.2
NP_001070937.1
5
723
NM_001077470.2
NP_001070938.1
4
583
NM_001077471.2
NP_001070939.1
5
723
NM_001077472.2
NP_001070940.1
4
583
NM_001077473.2
NP_001070941.1
4
583
NM_001077474.2
NP_001070942.1
5
723
NM_001077475.2
NP_001070943.1
4
583
NM_001077476.2
NP_001070944.1
4
583
NM_001077477.2
NP_001070945.1
4
583
NM_001077478.2
NP_001070946.1
5
723
NM_001077479.2
NP_001070947.1
4
583
NM_001077480.2
NP_001070948.1
5
723
NM_001077481.2
NP_001070949.1
5
723
NM_001077482.2
NP_001070950.1
5
723
NM_005122.4
NP_005113.1
5
723
XM_005245693.4
XP_005245750.1
4
996
XM_005245694.4
XP_005245751.1
4
995
XM_005245697.4
XP_005245754.1
5
606
XM_011510237.2
XP_011508539.1
4
993
GenBank mRNA
AB307702.1
-
4
520
AK303757.1
-
4
576
AK303796.1
-
5
757
AK310773.1
-
3
294
AK316208.1
-
5
715
AY572806.1
-
5
614
AY572807.1
-
5
543
AY572808.1
-
4
474
AY572809.1
-
3
304
AY572810.1
-
5
614
AY572811.1
-
4
474
AY572812.1
-
4
474
AY572813.1
-
5
614
AY572814.1
-
3
304
AY572815.1
-
3
304
AY572816.1
-
4
474
AY572817.1
-
4
474
AY572818.1
-
4
474
AY572819.1
-
5
614
AY572820.1
-
4
384
AY572826.1
-
4
444
AY572827.1
-
4
474
BC069626.1
-
5
676
7262456
BC121120.1
-
5
624
40120664
BC121121.1
-
5
624
40120665
DQ022681.1
-
4
520
HQ692838.1
-
4
520
HQ692839.1
-
4
520
HQ692840.1
-
4
520
HQ692841.1
-
4
520
HQ709178.1
-
4
520
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
transcription initiation from RNA polymerase II promoter
signal transduction
androgen receptor signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
androgen receptor activity
thyroid hormone receptor activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-xmxnb:80/100.66.76.46:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0