Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD36 SIRNA Assays ›
          Gene Symbol:
          ANKRD36
          Gene Name:
          ankyrin repeat domain 36
          Gene Aliases:
          UNQ2430
          Chromosome Location:
          Chr.2: 97113186 - 97264521 on Build GRCh38
          UniGene ID:
          Hs.541894
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s200612

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164315.1 NP_001157787.1 1 375
          XM_006712514.3 XP_006712577.1 1 685
          XM_011511130.1 XP_011509432.1 1 685
          XM_011511131.1 XP_011509433.1 1 685
          XM_017004009.1 XP_016859498.1 1 685
          XM_017004010.1 XP_016859499.1 1 685
          XM_017004011.1 XP_016859500.1 1 685
          XM_017004012.1 XP_016859501.1 1 685
          XM_017004013.1 XP_016859502.1 1 685
          XM_017004014.1 XP_016859503.1 1 685
          XM_017004015.1 XP_016859504.1 1 685
          XM_017004016.1 XP_016859505.1 1 685
          XM_017004017.1 XP_016859506.1 1 685
          XM_017004018.1 XP_016859507.1 1 685
          XM_017004019.1 XP_016859508.1 1 685
          XM_017004020.1 XP_016859509.1 1 685
          XM_017004021.1 XP_016859510.1 1 685
          XM_017004022.1 XP_016859511.1 1 685
          XM_017004023.1 XP_016859512.1 1 685
          XM_017004024.1 XP_016859513.1 1 685
          XM_017004025.1 XP_016859514.1 1 685
          XM_017004026.1 XP_016859515.1 1 685
          XM_017004027.1 XP_016859516.1 1 685
          XM_017004028.1 XP_016859517.1 1 685
          XM_017004029.1 XP_016859518.1 1 685
          XM_017004030.1 XP_016859519.1 1 685
          XM_017004031.1 XP_016859520.1 1 685
          XM_017004032.1 XP_016859521.1 1 685
          XM_017004033.1 XP_016859522.1 1 685
          XM_017004034.1 XP_016859523.1 1 685
          XM_017004035.1 XP_016859524.1 1 685
          XM_017004036.1 XP_016859525.1 1 685
          XM_017004037.1 XP_016859526.1 1 685
          XM_017004038.1 XP_016859527.1 1 685
          XM_017004039.1 XP_016859528.1 1 685
          XM_017004040.1 XP_016859529.1 1 685
          XM_017004041.1 XP_016859530.1 1 685
          XM_017004042.1 XP_016859531.1 1 685
          XM_017004043.1 XP_016859532.1 1 685
          XM_017004044.1 XP_016859533.1 1 685
          XM_017004045.1 XP_016859534.1 1 685
          XM_017004046.1 XP_016859535.1 1 685
          XM_017004047.1 XP_016859536.1 1 685
          XM_017004048.1 XP_016859537.1 1 685
          XM_017004049.1 XP_016859538.1 1 685
          GenBank mRNA AK304740.1 - 1 315
          AK307629.1 - 1 282
          AK316541.1 - 1 315
          AY358571.1 - 1 374
          BC046186.1 - 1 254
          BC128262.1 - 1 214
          BC128517.1 - 1 214 40118468
          BX647769.1 - 1 320

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0