Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008636.4 | NP_032662.3 | 4 | 835 | ||
GenBank mRNA | AK030284.1 | - | 4 | 814 | ||
AK150295.1 | - | 5 | 897 | |||
AY260849.1 | - | 3 | 689 | |||
AY260850.1 | - | 3 | 689 | |||
BC017679.1 | - | 4 | 797 | 4210829 | ||
X71327.1 | - | 4 | 785 |