Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNIP1 SIRNA Assays ›
Gene Symbol:
TNIP1
Gene Name:
TNFAIP3 interacting protein 1
Gene Aliases:
ABIN-1, NAF1, VAN, nip40-1
Chromosome Location:
Chr.5: 151029943 - 151087660 on Build GRCh38
UniGene ID:
Hs.355141
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20175
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001252385.1
NP_001239314.1
15
1778
NM_001252386.1
NP_001239315.1
14
1606
NM_001252390.1
NP_001239319.1
15
1643
NM_001252391.1
NP_001239320.1
15
1675
NM_001252392.1
NP_001239321.1
15
1675
NM_001252393.1
NP_001239322.1
15
1778
NM_001258454.1
NP_001245383.1
15
1778
NM_001258455.1
NP_001245384.1
14
1573
NM_001258456.1
NP_001245385.1
14
1573
NM_006058.4
NP_006049.3
15
1778
XM_005268355.1
XP_005268412.1
15
1691
XM_006714751.1
XP_006714814.1
15
1691
XM_006714752.2
XP_006714815.1
15
1691
XM_017008945.1
XP_016864434.1
15
1691
XM_017008946.1
XP_016864435.1
15
1657
XM_017008947.1
XP_016864436.1
16
2160
XM_017008948.1
XP_016864437.1
14
1952
XM_017008949.1
XP_016864438.1
15
1691
XM_017008950.1
XP_016864439.1
15
1691
XM_017008951.1
XP_016864440.1
14
1952
GenBank mRNA
AB177543.1
-
14
1537
AB177544.1
-
14
1537
AB252970.1
-
15
1647
AB252971.1
-
15
1647
AB252972.1
-
15
1768
AB252973.1
-
15
1768
AB252976.1
-
13
1401
AB252977.1
-
13
1401
AB252978.1
-
13
1401
AB252979.1
-
13
1401
AB252980.1
-
13
1401
AB252981.1
-
13
1401
AB252982.1
-
15
1647
AJ011895.1
-
15
1647
AJ011896.1
-
15
1647
AK074400.1
-
15
1709
AK290999.1
-
15
1644
AK298685.1
-
14
1600
AK299975.1
-
14
1538
AK303573.1
-
15
1728
AY012155.1
-
15
1794
BC012133.1
-
15
1684
4549542
BC014008.1
-
15
1625
4040118
BX640647.1
-
14
1527
D30755.2
-
14
1537
U39403.1
-
11
1133
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MyD88-dependent toll-like receptor signaling pathway
translation
proteolysis
defense response
inflammatory response
leukocyte cell-cell adhesion
glycoprotein biosynthetic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of viral genome replication
positive regulation of transcription from RNA polymerase II promoter
positive regulation of inflammatory response
negative regulation of ERK1 and ERK2 cascade
modulation by symbiont of host I-kappaB kinase/NF-kappaB cascade
thiol-dependent ubiquitin-specific protease activity
protein binding
mitogen-activated protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0