Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC2 SIRNA Assays ›
Gene Symbol:
TACC2
Gene Name:
transforming acidic coiled-coil containing protein 2
Gene Aliases:
AZU-1, ECTACC
Chromosome Location:
Chr.10: 121989174 - 122254545 on Build GRCh38
UniGene ID:
Hs.501252
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20765
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291876.1
NP_001278805.1
14, 15
8266
NM_001291877.1
NP_001278806.1
14, 15
8413
NM_001291878.1
NP_001278807.1
11, 12
2951
NM_001291879.1
NP_001278808.1
11, 12
2624
NM_006997.3
NP_008928.1
11, 12
3005
NM_206860.2
NP_996742.1
10, 11
2915
NM_206861.2
NP_996743.1
14, 15
3070
NM_206862.3
NP_996744.3
17, 18
8632
XM_005269388.1
XP_005269445.1
18, 19
8549
XM_005269389.4
XP_005269446.1
18, 19
9073
XM_005269390.1
XP_005269447.1
19, 20
8492
XM_005269391.1
XP_005269448.1
17, 18
8489
XM_005269392.1
XP_005269449.1
17, 18
8459
XM_005269393.1
XP_005269450.1
18, 19
8456
XM_005269394.1
XP_005269451.1
17, 18
8423
XM_005269395.1
XP_005269452.1
17, 18
8414
XM_005269396.1
XP_005269453.1
16, 17
8318
XM_005269397.1
XP_005269454.1
16, 17
8288
XM_005269399.1
XP_005269456.1
15, 16
8090
XM_006717548.1
XP_006717611.1
18, 19
8444
XM_006717552.1
XP_006717615.1
17, 18
8261
XM_006717553.1
XP_006717616.1
16, 17
8225
XM_006717554.1
XP_006717617.1
16, 17
8213
XM_011539126.1
XP_011537428.1
18, 19
8546
XM_011539128.1
XP_011537430.1
16, 17
3122
XM_011539129.1
XP_011537431.1
15, 16
2987
XM_011539130.1
XP_011537432.1
15, 16
2894
XM_011539131.1
XP_011537433.1
11, 12
2960
XM_011539132.1
XP_011537434.1
12, 13
2903
XM_011539133.1
XP_011537435.1
10, 11
2577
XM_011539134.1
XP_011537436.1
10, 11
2848
XM_017015484.1
XP_016870973.1
18, 19
8537
XM_017015485.1
XP_016870974.1
18, 19
8402
XM_017015486.1
XP_016870975.1
17, 18
8366
XM_017015487.1
XP_016870976.1
17, 18
8354
XM_017015488.1
XP_016870977.1
16, 17
8183
XM_017015489.1
XP_016870978.1
15, 16
8105
XM_017015491.1
XP_016870980.1
11, 12
3100
XM_017015492.1
XP_016870981.1
11, 12
3007
XM_017015493.1
XP_016870982.1
10, 11
2765
XM_017015494.1
XP_016870983.1
9, 10
2776
XM_017015495.1
XP_016870984.1
9, 10
2636
XM_017015496.1
XP_016870985.1
12, 13
2901
XM_017015497.1
XP_016870986.1
12, 13
3160
XM_017015498.1
XP_016870987.1
11, 12
3025
XM_017015499.1
XP_016870988.1
13, 14
2994
GenBank mRNA
AF095791.2
-
11, 12
2592
AF176646.1
-
10, 11
2750
AF220152.2
-
7, 8
2438
AF528098.1
-
14, 15
3051
AF528099.1
-
17, 18
8613
AK027066.1
-
7, 8
1544
AK075097.1
-
7, 8
902
AL713712.1
-
7, 8
1850
AL833304.1
-
16, 17
7731
BC039311.1
-
10, 11
2915
BC141837.1
-
12, 13
2603
BC142673.1
-
12, 13
2603
BC144599.1
-
14, 15
8381
9053129
BC144600.1
-
14, 15
8234
9053130
BC150627.1
-
14, 15
8234
9056997
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell proliferation
cerebral cortex development
nuclear hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0