Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RIPK2 SIRNA Assays ›
          Gene Symbol:
          RIPK2
          Gene Name:
          receptor interacting serine/threonine kinase 2
          Gene Aliases:
          CARD3, CARDIAK, CCK, GIG30, RICK, RIP2
          Chromosome Location:
          Chr.8: 89757747 - 89791064 on Build GRCh38
          UniGene ID:
          Hs.103755
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s225034

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RIPK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003821.5 NP_003812.1 3 770
          XM_005251092.3 XP_005251149.1 2 580
          GenBank mRNA AF027706.1 - 3 680
          AF064824.1 - 3 698
          AF078530.1 - 3 456
          AK075213.1 - 3 674
          AK301448.1 - 2 529
          AY358813.1 - 2 527
          AY358814.1 - 3 707
          AY524045.1 - 3 543
          AY562996.1 - 2 222
          BC004553.2 - 3 658 4026156
          KU178437.1 - 3 456
          KU178438.1 - 2 302

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          positive regulation of cytokine-mediated signaling pathway
          adaptive immune response
          apoptotic process
          inflammatory response
          signal transduction
          I-kappaB kinase/NF-kappaB signaling
          JNK cascade
          positive regulation of peptidyl-threonine phosphorylation
          peptidyl-tyrosine phosphorylation
          positive regulation of protein ubiquitination
          lipopolysaccharide-mediated signaling pathway
          positive regulation of chemokine production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-2 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          positive regulation of immature T cell proliferation
          positive regulation of peptidyl-serine phosphorylation
          toll-like receptor 2 signaling pathway
          toll-like receptor 4 signaling pathway
          T cell proliferation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to exogenous dsRNA
          innate immune response
          positive regulation of T-helper 1 cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          positive regulation of alpha-beta T cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-positive bacterium
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of ERK1 and ERK2 cascade
          nucleotide-binding oligomerization domain containing signaling pathway
          nucleotide-binding oligomerization domain containing 1 signaling pathway
          nucleotide-binding oligomerization domain containing 2 signaling pathway
          response to interleukin-1
          response to interleukin-12
          response to interleukin-18
          cellular response to lipoteichoic acid
          cellular response to peptidoglycan
          cellular response to muramyl dipeptide
          positive regulation of xenophagy
          protein serine/threonine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signal transducer activity
          receptor binding
          protein binding
          ATP binding
          LIM domain binding
          protein homodimerization activity
          CARD domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jct7h:80/100.66.72.173:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0