Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PMS1 SIRNA Assays ›
Gene Symbol:
PMS1
Gene Name:
PMS1 homolog 1, mismatch repair system component
Gene Aliases:
HNPCC3, MLH2, PMSL1, hPMS1
Chromosome Location:
Chr.2: 189784085 - 189877629 on Build GRCh38
UniGene ID:
Hs.111749
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s229951
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PMS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000534.4
NP_000525.1
9
1890
NM_001128143.1
NP_001121615.1
8
1773
NM_001128144.1
NP_001121616.1
9
1890
NM_001289408.1
NP_001276337.1
8
1787
NM_001289409.1
NP_001276338.1
7
1604
NM_001321044.1
NP_001307973.1
8
1773
NM_001321045.1
NP_001307974.1
10
2016
NM_001321046.1
NP_001307975.1
8
1707
NM_001321047.1
NP_001307976.1
9
2067
NM_001321048.1
NP_001307977.1
10
1987
NR_110332.1
9
1870
XM_006712596.1
XP_006712659.1
5
855
XM_011511356.2
XP_011509658.1
9
2176
XM_017004344.1
XP_016859833.1
9
1499
XM_017004345.1
XP_016859834.1
8
1324
XM_017004346.1
XP_016859835.1
8
1287
XM_017004347.1
XP_016859836.1
7
1190
XM_017004348.1
XP_016859837.1
9
1616
XM_017004349.1
XP_016859838.1
8
2059
XM_017004350.1
XP_016859839.1
8
1316
XM_017004351.1
XP_016859840.1
7
1278
GenBank mRNA
AB102869.1
-
7
1244
AB102870.1
-
8
1361
AB102872.1
-
4
716
AB102875.1
-
7
1244
AK295602.1
-
8
1383
AK297440.1
-
8
1330
AK304634.1
-
8
1226
AK316215.1
-
7
1124
AY540750.1
-
4
879
AY540751.1
-
9
1442
BC036376.1
-
9
1609
BC096330.1
-
9
1431
BC096331.1
-
9
1430
40008290
BC096332.1
-
9
1431
BM800196.1
-
3
598
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
mismatch repair
DNA binding
single-stranded DNA binding
ATP binding
ATPase activity
mismatched DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0