Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHLDA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012396.4 | NP_036528.1 | 2 | 1333 | ||
NR_073080.1 | 3 | 1427 | ||||
GenBank mRNA | AF151100.1 | - | 2 | 1264 | ||
AK075179.1 | - | 2 | 1296 | |||
AK222644.1 | - | 2 | 1296 | |||
BC014390.1 | - | 2 | 1185 | 3547186 | ||
BC068273.1 | - | 2 | 1333 | 5177273 |