Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281453.1 | NP_001268382.1 | 3 | 470 | ||
NM_001281454.1 | NP_001268383.1 | 3 | 374 | |||
GenBank mRNA | AF072247.1 | - | 3 | 363 | ||
AK001474.1 | - | 2 | 170 | |||
AK021532.1 | - | 2 | 124 | |||
AK290882.1 | - | 3 | 417 | |||
BC009372.1 | - | 3 | 306 | 4122128 | ||
BC043619.1 | - | 3 | 324 | 6156267 | ||
BU181763.1 | - | 3 | 470 |