Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NLGN3 SIRNA Assays ›
          Gene Symbol:
          NLGN3
          Gene Name:
          neuroligin 3
          Gene Aliases:
          HNL3
          Chromosome Location:
          Chr.X: 71144389 - 71175307 on Build GRCh38
          UniGene ID:
          Hs.438877
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s28945

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NLGN3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166660.1 NP_001160132.1 5 1504
          NM_001321276.1 NP_001308205.1 6 1051
          NM_018977.3 NP_061850.2 6 1564
          NM_181303.1 NP_851820.1 7 1624
          XM_006724662.3 XP_006724725.2 8 1439
          XM_006724663.3 XP_006724726.1 8 1631
          XM_011530974.2 XP_011529276.1 7 1553
          XM_017029597.1 XP_016885086.1 6 1426
          GenBank mRNA AB040913.1 - 4 794
          AF217411.1 - 6 1525
          AF217412.1 - 6 1336
          AK296217.1 - 4 859
          AK299669.1 - 6 1035
          AK314699.1 - 5 1341
          BC028738.1 - 5 1499
          BC051715.1 - 6 1494 5258980
          BX641059.1 - 6 1047
          GQ489207.1 - 6 1290
          KJ534889.1 - 5 1230
          KJ534890.1 - 4 1170
          KJ534891.1 - 6 1290
          KJ534977.1 - 4 1170
          KJ535040.1 - 4 1170
          KU178651.1 - 5 1230

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of respiratory gaseous exchange by neurological system process
          receptor-mediated endocytosis
          neuron cell-cell adhesion
          synapse assembly
          learning
          visual learning
          adult behavior
          social behavior
          axon extension
          oligodendrocyte differentiation
          modulation of synaptic transmission
          synapse organization
          positive regulation of synapse assembly
          positive regulation of synaptic transmission, glutamatergic
          rhythmic synaptic transmission
          excitatory postsynaptic potential
          inhibitory postsynaptic potential
          long-term synaptic potentiation
          positive regulation of dendritic spine development
          negative regulation of dendritic spine morphogenesis
          vocalization behavior
          negative regulation of excitatory postsynaptic potential
          postsynaptic membrane assembly
          presynaptic membrane assembly
          positive regulation of inhibitory postsynaptic potential
          regulation of long-term synaptic potentiation
          positive regulation of protein localization to synapse
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          regulation of terminal button organization
          positive regulation of excitatory postsynaptic potential
          positive regulation of synaptic vesicle clustering
          positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          receptor activity
          protein binding
          neurexin family protein binding
          cell adhesion molecule binding
          carboxylic ester hydrolase activity
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0