Search
Search

BTNL2| Gene Symbol: BTNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304561.1 | NP_001291490.1 | 6 | 1196 | ||
| XM_017011057.1 | XP_016866546.1 | 6 | 2945 | |||
| GenBank mRNA | BC119668.1 | - | 5 | 666 | ||
| BC127641.1 | - | 5 | 667 | |||
| BC127642.1 | - | 4 | 559 | 40116164 | ||