Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA010727.1 | - | 3 | 549 |
Gene Symbol: KYAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008661.2 | NP_001008661.1 | 4 | 658 | ||
NM_001008662.2 | NP_001008662.1 | 3 | 558 | |||
GenBank mRNA | AK057176.1 | - | 3 | 374 | ||
AK301326.1 | - | 4 | 345 | |||
AK315917.1 | - | 4 | 355 | |||
AL833593.1 | - | 4 | 303 | |||
AY028624.1 | - | 4 | 478 | |||
BC000819.1 | - | 1 | 31 | 3453829 | ||
BM728982.1 | - | 4 | 620 | |||
CR627392.1 | - | 4 | 320 | |||
DC424120.1 | - | 4 | 661 |