Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PARD3 SIRNA Assays ›
          Gene Symbol:
          PARD3
          Gene Name:
          par-3 family cell polarity regulator
          Gene Aliases:
          ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2
          Chromosome Location:
          Chr.10: 34109560 - 34815325 on Build GRCh38
          UniGene ID:
          Hs.131489
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s32126

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PARD3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001184785.1 NP_001171714.1 15 2459
          NM_001184786.1 NP_001171715.1 14 2420
          NM_001184787.1 NP_001171716.1 15 2459
          NM_001184788.1 NP_001171717.1 14 2420
          NM_001184789.1 NP_001171718.1 14 2420
          NM_001184790.1 NP_001171719.1 13 2288
          NM_001184791.1 NP_001171720.1 13 2288
          NM_001184792.1 NP_001171721.1 15 2459
          NM_001184793.1 NP_001171722.1 15 2459
          NM_001184794.1 NP_001171723.1 14 2420
          NM_019619.3 NP_062565.2 15 2459
          XM_005252528.4 XP_005252585.1 15 2450
          XM_005252530.4 XP_005252587.1 15 2450
          XM_005252531.4 XP_005252588.1 15 2450
          XM_005252532.4 XP_005252589.1 14 2104
          XM_005252534.4 XP_005252591.1 15 2450
          XM_005252535.4 XP_005252592.1 14 2411
          XM_005252536.4 XP_005252593.1 15 2450
          XM_011519569.2 XP_011517871.1 15 2450
          XM_011519570.2 XP_011517872.1 14 2411
          XM_011519571.2 XP_011517873.1 15 2450
          XM_011519572.2 XP_011517874.1 15 2450
          XM_011519573.2 XP_011517875.1 15 2450
          XM_011519575.2 XP_011517877.1 14 2134
          XM_011519576.2 XP_011517878.1 14 2105
          XM_011519577.2 XP_011517879.1 13 2282
          XM_011519578.2 XP_011517880.1 13 2283
          XM_011519579.2 XP_011517881.1 13 2003
          XM_011519581.2 XP_011517883.1 15 2450
          XM_011519583.2 XP_011517885.1 15 2450
          XM_011519584.2 XP_011517886.1 15 2450
          XM_017016430.1 XP_016871919.1 15 2450
          GenBank mRNA AA136967.1 - 1 17
          AB073671.1 - 13 1958
          AF177228.1 - 8 1195
          AF196185.1 - 15 2416
          AF196186.1 - 15 2416
          AF252293.1 - 13 1958
          AF332592.1 - 14 2090
          AF332593.1 - 14 2090
          AF454057.1 - 13 1836
          AF454058.1 - 13 2302
          AF454059.1 - 10 1423
          AF467002.1 - 15 2129
          AF467003.1 - 15 2129
          AF467004.1 - 15 2129
          AF467005.1 - 13 1958
          AF467006.1 - 14 2090
          AK024668.1 - 2 111
          AK025892.1 - 6 906
          AK027735.1 - 13 1756
          BC071566.1 - 14 2367 30344732

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          protein targeting to membrane
          cell cycle
          establishment or maintenance of cell polarity
          transforming growth factor beta receptor signaling pathway
          protein kinase C-activating G-protein coupled receptor signaling pathway
          axonogenesis
          asymmetric cell division
          negative regulation of peptidyl-threonine phosphorylation
          myelination in peripheral nervous system
          positive regulation of myelination
          regulation of cellular localization
          bicellular tight junction assembly
          establishment of epithelial cell polarity
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          phosphatidylinositol-3,4,5-trisphosphate binding
          protein phosphatase binding
          phosphatidylinositol-3-phosphate binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nq57x:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0