Search
Search

ZNF319| Gene Symbol: ZNF319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020807.2 | NP_065858.1 | 2 | 2208 | ||
| XM_005256069.3 | XP_005256126.1 | 2 | 2133 | |||
| GenBank mRNA | AB037809.1 | - | 2 | 2208 | ||
| BC093919.1 | - | 1 | 1647 | 7939764 | ||
| BC101767.1 | - | 1 | 1647 | 8069273 | ||