Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286274.1 | NP_001273203.1 | 3 | 464 | ||
NM_001286275.1 | NP_001273204.1 | 4 | 523 | |||
NM_001286276.1 | NP_001273205.1 | 4 | 475 | |||
NM_020931.3 | NP_065982.1 | 4 | 545 | |||
XM_011514767.2 | XP_011513069.1 | 4 | 386 | |||
GenBank mRNA | AB046806.1 | - | 1 | 1657 | ||
AK290988.1 | - | 4 | 339 | |||
AK295519.1 | - | 3 | 270 | |||
BC029372.1 | - | 4 | 354 | |||
BC041125.1 | - | 4 | 491 | 5785266 | ||
BF699031.1 | - | 4 | 472 | |||
BG028983.1 | - | 311 |