Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM62 SIRNA Assays ›
Gene Symbol:
TMEM62
Gene Name:
transmembrane protein 62
Gene Aliases:
-
Chromosome Location:
Chr.15: 43133115 - 43185147 on Build GRCh38
UniGene ID:
Hs.511175
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36839
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM62
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024956.3
NP_079232.3
11
1647
XM_005254682.4
XP_005254739.1
10
1614
XM_005254683.4
XP_005254740.1
9
1503
XM_005254684.3
XP_005254741.1
8
1360
XM_011522071.1
XP_011520373.1
10
1131
XM_011522072.2
XP_011520374.1
9
1298
XM_011522073.1
XP_011520375.1
7
796
XM_011522074.2
XP_011520376.1
10
1442
XM_017022600.1
XP_016878089.1
10
1646
XM_017022601.1
XP_016878090.1
10
1399
XM_017022602.1
XP_016878091.1
9
1535
XM_017022603.1
XP_016878092.1
10
1131
XM_017022604.1
XP_016878093.1
10
1615
XM_017022605.1
XP_016878094.1
9
1504
XM_017022606.1
XP_016878095.1
9
1472
XM_017022607.1
XP_016878096.1
8
1392
XM_017022608.1
XP_016878097.1
8
926
XM_017022609.1
XP_016878098.1
9
1473
XM_017022610.1
XP_016878099.1
9
1233
XM_017022611.1
XP_016878100.1
9
1288
XM_017022612.1
XP_016878101.1
9
1407
XM_017022613.1
XP_016878102.1
8
1099
XM_017022614.1
XP_016878103.1
8
1057
XM_017022615.1
XP_016878104.1
8
1038
XM_017022616.1
XP_016878105.1
8
1096
GenBank mRNA
AK027028.1
-
8
952
AK225353.1
-
8
952
AK309679.1
-
10
1163
BC009981.2
-
11
1647
4121449
CR457370.1
-
6
744
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA helicase
Gene Ontology Categories:
Process(es)
hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0