Search
Search

ORAI2| Gene Symbol: ORAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126340.2 | NP_001119812.1 | 4 | 1410 | ||
| NM_001271818.1 | NP_001258747.1 | 4 | 1433 | |||
| NM_001271819.1 | NP_001258748.1 | 3 | 1172 | |||
| NM_032831.3 | NP_116220.1 | 3 | 1301 | |||
| GenBank mRNA | AF258552.1 | - | 1 | 588 | ||
| AK022536.1 | - | 1 | 242 | |||
| AK027639.1 | - | 3 | 1255 | |||
| AK056497.1 | - | 4 | 1365 | |||
| BC069270.1 | - | 3 | 1204 | 5477442 | ||
| CR457287.1 | - | 1 | 217 | |||
| Y10777.1 | - | 1 | 371 | |||