Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREB3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052854.3 | NP_443086.1 | 9 | 1540 | ||
GenBank mRNA | AB063321.1 | - | 9 | 1161 | ||
AF055009.1 | - | 1 | 660 | |||
AK075538.1 | - | 4 | 311 | |||
AK314072.1 | - | 9 | 1518 | |||
BC014097.1 | - | 9 | 1183 | 4546950 | ||
BC015781.1 | - | 9 | 1169 | 4856946 |