Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DYNC1I2 SIRNA Assays ›
Gene Symbol:
DYNC1I2
Gene Name:
dynein cytoplasmic 1 intermediate chain 2
Gene Aliases:
DIC74, DNCI2, IC2
Chromosome Location:
Chr.2: 171687409 - 171750158 on Build GRCh38
UniGene ID:
Hs.546250
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4206
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DYNC1I2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271785.1
NP_001258714.1
17
1994
NM_001271786.1
NP_001258715.1
15
1987
NM_001271787.1
NP_001258716.1
15
1970
NM_001271788.1
NP_001258717.1
15
1933
NM_001271789.1
NP_001258718.1
15
1916
NM_001271790.1
NP_001258719.1
15
1933
NM_001320882.1
NP_001307811.1
16
1993
NM_001320883.1
NP_001307812.1
16
1976
NM_001320884.1
NP_001307813.1
15
1916
NM_001378.2
NP_001369.1
17
2011
XM_006712347.3
XP_006712410.1
16
1917
XM_017003525.1
XP_016859014.1
16
1952
GenBank mRNA
AB209014.1
-
13
1936
AF134477.1
-
15
1805
AF250307.1
-
15
1825
AK024742.1
-
1
318
AK026574.1
-
1
1019
AK055491.1
-
17
1948
AK096579.1
-
17
1912
AK225759.1
-
15
1879
AK298561.1
-
15
1830
AK301866.1
-
7
1146
AK316119.1
-
15
1850
AL137519.1
-
9
1156
AL832832.1
-
11
1303
AY037160.1
-
15
1841
BC015038.2
-
15
1777
3921299
BC091498.1
-
15
1861
30386904
BC109375.1
-
15
1818
4608676
BT007130.1
-
14
1705
BX537412.1
-
15
1873
DQ786313.1
-
2
155
DQ786324.1
-
11
1331
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00398527_m1
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
ER to Golgi vesicle-mediated transport
microtubule-based movement
viral process
antigen processing and presentation of exogenous peptide antigen via MHC class II
microtubule motor activity
protein binding
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0