Search
Search

SLC47A2| Gene Symbol: SLC47A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099646.1 | NP_001093116.1 | 10, 11 | 1082 | ||
| NM_152908.3 | NP_690872.2 | 10, 11 | 1190 | |||
| NR_135624.1 | 10, 11 | 1653 | ||||
| NR_135625.1 | 10, 11 | 1236 | ||||
| GenBank mRNA | AB250364.1 | - | 10, 11 | 925 | ||
| AB250701.1 | - | 10, 11 | 1079 | |||
| AK055758.1 | - | 10, 11 | 964 | |||
| AK307647.1 | - | 10, 11 | 1082 | |||
| AK310218.1 | - | 8, 9 | 796 | |||
| BC050578.1 | - | 10, 11 | 1066 | 5759988 | ||
| BG432484.1 | - | 2, 3 | 167 | |||
| BM924669.1 | - | 918 | ||||
| BX648861.1 | - | 10, 11 | 1653 | |||