Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CNTN4 SIRNA Assays ›
          Gene Symbol:
          CNTN4
          Gene Name:
          contactin 4
          Gene Aliases:
          AXCAM, BIG-2
          Chromosome Location:
          Chr.3: 2098771 - 3059080 on Build GRCh38
          UniGene ID:
          Hs.298705
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s45685

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CNTN4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206955.1 NP_001193884.1 19 2665
          NM_001206956.1 NP_001193885.1 11 1384
          NM_175607.2 NP_783200.1 20 2620
          NM_175613.2 NP_783302.1 11 1387
          XM_006713004.3 XP_006713067.1 19 2630
          XM_011533425.2 XP_011531727.1 19 2633
          XM_011533426.2 XP_011531728.1 20 2852
          XM_011533427.2 XP_011531729.1 20 2833
          XM_011533428.2 XP_011531730.1 18 2822
          XM_011533429.2 XP_011531731.1 18 2757
          XM_011533430.2 XP_011531732.1 18 2718
          XM_011533431.2 XP_011531733.1 17 5467
          XM_017005782.1 XP_016861271.1 20 2575
          XM_017005783.1 XP_016861272.1 21 3449
          XM_017005784.1 XP_016861273.1 19 2910
          XM_017005785.1 XP_016861274.1 20 2557
          XM_017005786.1 XP_016861275.1 18 2819
          XM_017005787.1 XP_016861276.1 18 2715
          GenBank mRNA AF464063.1 - 1 531
          AF549455.1 - 17 2663
          AK057278.1 - 11 1365
          AK097633.1 - 14 1911
          AK125460.1 - 19 2808
          AK314396.1 - 11 1365
          AY090737.1 - 19 2502
          BC026119.1 - 11 1367 4826091

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          neuron cell-cell adhesion
          nervous system development
          axonogenesis
          axon guidance
          axonal fasciculation
          brain development
          neuron projection development
          negative regulation of neuron differentiation
          regulation of synaptic plasticity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hpnss:80/100.66.78.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0