Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELN SIRNA Assays ›
Gene Symbol:
ELN
Gene Name:
elastin
Gene Aliases:
SVAS, WBS, WS
Chromosome Location:
Chr.7: 74027772 - 74069907 on Build GRCh38
UniGene ID:
Hs.647061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4641
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000501.3
NP_000492.2
33
3516
NM_001081752.2
NP_001075221.1
30
3375
NM_001081753.2
NP_001075222.1
31
3420
NM_001081754.2
NP_001075223.1
32
3477
NM_001081755.2
NP_001075224.1
32
3459
NM_001278912.1
NP_001265841.1
32
3462
NM_001278913.1
NP_001265842.1
29
3273
NM_001278914.1
NP_001265843.1
31
3444
NM_001278915.1
NP_001265844.1
33
3534
NM_001278916.1
NP_001265845.1
29
3318
NM_001278917.1
NP_001265846.1
32
3486
NM_001278918.1
NP_001265847.1
28
3195
NM_001278939.1
NP_001265868.1
34
3702
XM_005250187.1
XP_005250244.1
32
3157
XM_005250188.1
XP_005250245.1
32
3149
XM_011515868.1
XP_011514170.1
33
3217
XM_011515869.1
XP_011514171.1
32
3146
XM_011515870.1
XP_011514172.1
32
3512
XM_011515871.1
XP_011514173.1
32
3175
XM_011515872.1
XP_011514174.1
32
3163
XM_011515873.1
XP_011514175.1
32
3160
XM_011515874.1
XP_011514176.1
31
3482
XM_011515875.1
XP_011514177.1
31
3138
XM_011515876.1
XP_011514178.1
31
3118
XM_011515877.1
XP_011514179.1
31
3106
XM_017011813.1
XP_016867302.1
31
3060
XM_017011814.1
XP_016867303.1
31
3118
GenBank mRNA
AB208942.1
-
32
3112
AF086171.1
-
1
179
AK075494.1
-
15
1458
AK075554.1
-
29
2957
AK092232.1
-
28
2810
AK094138.1
-
33
2949
AK095990.1
-
31
3040
AK122731.1
-
25
2669
AK125511.1
-
31
3102
AK130894.1
-
18
2011
AK225659.1
-
32
3073
BC035570.1
-
33
3121
BC050379.1
-
26
2390
BC065566.1
-
29
2950
6153564
BX537939.1
-
32
3097
BX538199.1
-
30
3007
Back To Top
More Information
Panther Classification:
Molecular Function -
structural protein
Gene Ontology Categories:
Function(s)
Process(es)
respiratory gaseous exchange
blood circulation
cell proliferation
organ morphogenesis
extracellular matrix disassembly
extracellular matrix organization
extracellular matrix structural constituent
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0