Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FLCN SIRNA Assays ›
          Gene Symbol:
          FLCN
          Gene Name:
          folliculin
          Gene Aliases:
          BHD, FLCL
          Chromosome Location:
          Chr.17: 17206945 - 17237188 on Build GRCh38
          UniGene ID:
          Hs.31652
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s529110

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FLCN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_144606.5 NP_653207.1 7 1234
          NM_144997.5 NP_659434.2 7 1234
          XM_011523714.2 XP_011522016.1 8 1285
          XM_011523716.1 XP_011522018.1 6 1663
          XM_011523718.2 XP_011522020.1 7 1169
          XM_011523719.2 XP_011522021.1 8 1285
          XM_011523721.2 XP_011522023.1 7 1034
          XM_017024304.1 XP_016879793.1 9 1454
          XM_017024305.1 XP_016879794.1 9 1469
          XM_017024306.1 XP_016879795.1 8 1458
          XM_017024307.1 XP_016879796.1 8 1420
          XM_017024308.1 XP_016879797.1 9 1596
          XM_017024309.1 XP_016879798.1 6 1009
          GenBank mRNA AF517523.1 - 7 1185
          AL831885.1 - 1 573
          BC015687.2 - 7 1185 4684974
          KU178888.1 - 4 730
          KU178889.1 - 3 508

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          cell-cell junction assembly
          positive regulation of autophagy
          negative regulation of gene expression
          negative regulation of mitochondrion organization
          hemopoiesis
          negative regulation of cell growth
          negative regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          TOR signaling
          regulation of TOR signaling
          negative regulation of TOR signaling
          positive regulation of TOR signaling
          regulation of cytokinesis
          negative regulation of Rho protein signal transduction
          regulation of histone acetylation
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          positive regulation of cell adhesion
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of protein kinase B signaling
          negative regulation of ERK1 and ERK2 cascade
          negative regulation of protein localization to nucleus
          negative regulation of cell proliferation involved in kidney development
          negative regulation of energy homeostasis
          regulation of pro-B cell differentiation
          negative regulation of ATP biosynthetic process
          guanyl-nucleotide exchange factor activity
          protein binding
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0