Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC392196 SIRNA Assays ›
          Gene Symbol:
          LOC392196 USP17L1 USP17L2 USP17L3 USP17L4 USP17L7 USP17L8
          Gene Name:
          ubiquitin specific peptidase 17-like family member 2 pseudogene
          ubiquitin specific peptidase 17-like family member 1
          ubiquitin specific peptidase 17-like family member 2
          ubiquitin specific peptidase 17-like family member 3
          ubiquitin specific peptidase 17-like family member 4
          ubiquitin specific peptidase 17-like family member 7
          ubiquitin specific peptidase 17-like family member 8
          Gene Aliases:
          -
          USP17L1P
          DUB-3, DUB3, USP17
          USP17B, USP17F
          -
          -
          USP17L10
          Chromosome Location:
          Chr.8: 12127858 - 12129297 on Build GRCh38
          Chromosome Location:
          Chr.8: 7332387 - 7333979 on Build GRCh38
          Chromosome Location:
          Chr.8: 12137168 - 12138760 on Build GRCh38
          Chromosome Location:
          Chr.8: 7976393 - 7977985 on Build GRCh38
          Chromosome Location:
          Chr.8: 7337115 - 7338707 on Build GRCh38
          Chromosome Location:
          Chr.8: 12132417 - 12134009 on Build GRCh38
          Chromosome Location:
          Chr.8: 7971661 - 7973253 on Build GRCh38
          UniGene ID:
          Hs.742129
          Hs.653091
          Hs.531448
          Hs.741131
          Hs.741134
          Hs.741132
          Hs.741135
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s53035

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC392196
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003275.2 1 1103
          GenBank mRNA BF947332.1 - 1 276
          Gene Symbol: USP17L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256873.1 NP_001243802.1 1 1103
          Gene Symbol: USP17L2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_201402.2 NP_958804.2 1 1103
          Gene Symbol: USP17L3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256871.1 NP_001243800.1 1 1103
          Gene Symbol: USP17L4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256874.1 NP_001243803.1 1 1103
          Gene Symbol: USP17L7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256869.1 NP_001243798.1 1 1103
          Gene Symbol: USP17L8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256872.1 NP_001243801.1 1 1103

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          apoptotic process
          protein deubiquitination
          cell cycle checkpoint
          mitotic cell cycle checkpoint
          negative regulation of protein processing
          regulation of cell migration
          negative regulation of histone deacetylation
          negative regulation of GTPase activity
          regulation of cell proliferation
          regulation of apoptotic process
          positive regulation of GTPase activity
          regulation of defense response to virus by host
          protein K63-linked deubiquitination
          protein K48-linked deubiquitination
          CAAX-box protein processing
          negative regulation of protein targeting to membrane
          regulation of ruffle assembly
          positive regulation of MDA-5 signaling pathway
          positive regulation of RIG-I signaling pathway
          thiol-dependent ubiquitinyl hydrolase activity
          thiol-dependent ubiquitin-specific protease activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0