Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC8 SIRNA Assays ›
          Gene Symbol:
          HDAC8
          Gene Name:
          histone deacetylase 8
          Gene Aliases:
          CDA07, CDLS5, HD8, HDACL1, MRXS6, RPD3, WTS
          Chromosome Location:
          Chr.X: 72329516 - 72573103 on Build GRCh38
          UniGene ID:
          Hs.310536
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531276

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HDAC8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166419.1 NP_001159891.1 4 739
          NM_001166420.1 NP_001159892.1 4 739
          NM_001166422.1 NP_001159894.1 4 739
          NM_018486.2 NP_060956.1 4 739
          XM_011530986.2 XP_011529288.2 4 734
          XM_011530987.2 XP_011529289.2 4 734
          XM_017029640.1 XP_016885129.1 4 734
          XM_017029641.1 XP_016885130.1 4 734
          XM_017029643.1 XP_016885132.1 4 734
          XM_017029645.1 XP_016885134.1 4 734
          XM_017029646.1 XP_016885135.1 3 283
          XM_017029647.1 XP_016885136.1 4 734
          GenBank mRNA AF212246.1 - 4 455
          AF230097.1 - 4 452
          AF245664.1 - 4 406
          AJ277724.1 - 4 439
          AK074326.1 - 4 684
          AK300895.1 - 4 436
          AY429539.1 - 3 283
          BC050433.1 - 4 430 5761745
          BQ189619.1 - 4 489

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: AY066003.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          chromatin assembly or disassembly
          transcription, DNA-templated
          sister chromatid cohesion
          chromatin modification
          negative regulation of protein ubiquitination
          regulation of protein stability
          regulation of telomere maintenance
          histone H3 deacetylation
          regulation of cohesin loading
          histone deacetylase activity
          protein binding
          transcription factor binding
          Hsp70 protein binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          metal ion binding
          Hsp90 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0