Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STAMBP SIRNA Assays ›
          Gene Symbol:
          STAMBP
          Gene Name:
          STAM binding protein
          Gene Aliases:
          AMSH, MICCAP
          Chromosome Location:
          Chr.2: 73828911 - 73873656 on Build GRCh38
          UniGene ID:
          Hs.469018
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s532094

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STAMBP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006463.4 NP_006454.1 3, 4 411
          NM_201647.2 NP_964010.1 2, 3 302
          NM_213622.2 NP_998787.1 2, 3 734
          XM_005264088.3 XP_005264145.1 2, 3 309
          XM_011532483.2 XP_011530785.1 3, 4 406
          XM_011532484.2 XP_011530786.1 3, 4 424
          XM_011532485.2 XP_011530787.1 3, 4 406
          XM_017003175.1 XP_016858664.1 3, 4 357
          XM_017003176.1 XP_016858665.1 2, 3 307
          XM_017003177.1 XP_016858666.1 3, 4 412
          XM_017003178.1 XP_016858667.1 2, 3 760
          XM_017003179.1 XP_016858668.1 3, 4 358
          XM_017003180.1 XP_016858669.1 2, 3 291
          XM_017003181.1 XP_016858670.1 2, 3 749
          GenBank mRNA AF052135.1 - 2, 3 325
          BC007682.1 - 1, 2 225 3619837
          BC065574.1 - 2, 3 712 6170296
          BC101467.1 - 2, 3 300 8068973
          BC101469.1 - 2, 3 298 8068975
          BF203378.1 - 394
          BM478027.1 - 3, 4 398
          BQ929694.1 - 2, 3 260
          CX758449.1 - 2, 3 733
          DB477012.1 - 3, 4 412
          DN919763.1 - 2, 3 317
          DR158757.1 - 3, 4 396
          EU927390.1 - 1, 2 204
          U73522.1 - 1, 2 219

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          JAK-STAT cascade
          positive regulation of cell proliferation
          negative regulation of phosphatidylinositol 3-kinase signaling
          protein deubiquitination
          negative regulation of neuron apoptotic process
          negative regulation of Ras protein signal transduction
          thiol-dependent ubiquitin-specific protease activity
          protein binding
          metallopeptidase activity
          protein domain specific binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0