Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFP36L1 SIRNA Assays ›
Gene Symbol:
ZFP36L1
Gene Name:
ZFP36 ring finger protein like 1
Gene Aliases:
BRF1, Berg36, ERF-1, ERF1, RNF162B, TIS11B, cMG1
Chromosome Location:
Chr.14: 68787655 - 68796243 on Build GRCh38
UniGene ID:
Hs.707091
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s53256
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFP36L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001244701.1
NP_001231630.1
1
46
GenBank mRNA
DA633985.1
-
1
46
FY210721.1
-
1
17
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
vasculogenesis
proepicardium development
regulation of transcription, DNA-templated
apoptotic process
heart development
cell proliferation
response to wounding
regulation of gene expression
regulation of keratinocyte proliferation
phosphatidylinositol 3-kinase signaling
neural tube development
nuclear-transcribed mRNA catabolic process, deadenylation-independent decay
regulation of mRNA 3'-end processing
cellular response to insulin stimulus
T cell differentiation in thymus
multicellular organism growth
p38MAPK cascade
regulation of mRNA stability
protein kinase B signaling
cellular response to fibroblast growth factor stimulus
positive regulation of fat cell differentiation
regulation of keratinocyte differentiation
negative regulation of erythrocyte differentiation
positive regulation of monocyte differentiation
regulation of myoblast differentiation
mesendoderm development
mRNA transport
chorio-allantoic fusion
spongiotrophoblast layer development
3'-UTR-mediated mRNA destabilization
ERK1 and ERK2 cascade
cellular response to cAMP
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
cellular response to peptide hormone stimulus
cellular response to glucocorticoid stimulus
cellular response to hypoxia
cellular response to salt stress
cellular response to transforming growth factor beta stimulus
regulation of stem cell proliferation
cellular response to raffinose
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
regulation of keratinocyte apoptotic process
positive regulation of intracellular mRNA localization
cellular response to phorbol 13-acetate 12-myristate
DNA binding
transcription factor activity, sequence-specific DNA binding
mRNA binding
mRNA 3'-UTR binding
protein binding
mRNA 3'-UTR AU-rich region binding
poly(A) RNA binding
metal ion binding
14-3-3 protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0