Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPIPB8 SIRNA Assays ›
          Gene Symbol:
          NPIPB8 NPIPB15 NPIPB6 NPIPB9 PDXDC2P
          Gene Name:
          nuclear pore complex interacting protein family member B8
          nuclear pore complex interacting protein family member B15
          nuclear pore complex interacting protein family member B6
          nuclear pore complex interacting protein family member B9
          pyridoxal dependent decarboxylase domain containing 2, pseudogene
          Gene Aliases:
          -
          A-761H5.4, NPIPL2
          NPIPB
          -
          NPIP, PDXDC2
          Chromosome Location:
          Chr.16: 28638065 - 28658693 on Build GRCh38
          Chromosome Location:
          Chr.16: 74374986 - 74392115 on Build GRCh38
          Chromosome Location:
          Chr.16: 28342517 - 28364971 on Build GRCh38
          Chromosome Location:
          Chr.16: 28735500 - 28772911 on Build GRCh38
          Chromosome Location:
          Chr.16: 69976297 - 70065948 on Build GRCh38
          UniGene ID:
          -
          -
          Hs.726467
          Hs.710214
          Hs.513695
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s54088

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPIPB8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310136.1 NP_001297065.1 7 1137
          XM_017023622.1 XP_016879111.1 8 1408
          XM_017023623.1 XP_016879112.1 9 1345
          XM_017023624.1 XP_016879113.1 8 1261
          XM_017023625.1 XP_016879114.1 8 1241
          XM_017023626.1 XP_016879115.1 9 1880
          Gene Symbol: NPIPB15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001306094.1 NP_001293023.1 7 1090
          XM_011523496.2 XP_011521798.1 8 1146
          XM_011523498.2 XP_011521800.1 8 1180
          XM_011523501.2 XP_011521803.1 4 972
          XM_017023248.1 XP_016878737.1 8 1583
          XM_017023249.1 XP_016878738.1 7 1400
          Gene Symbol: NPIPB6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282524.1 NP_001269453.1 7 1112
          XM_005255741.3 XP_005255798.1 8 1924
          XM_005255743.3 XP_005255800.1 4 2503
          XM_011545929.2 XP_011544231.1 9 1744
          XM_011545931.2 XP_011544233.1 8 1377
          XM_017023628.1 XP_016879117.1 8 1127
          XM_017023629.1 XP_016879118.1 8 1670
          XM_017023630.1 XP_016879119.1 9 1397
          XM_017023631.1 XP_016879120.1 7 1741
          XM_017023632.1 XP_016879121.1 8 1127
          GenBank mRNA BC144439.1 - 7 1073
          BC157836.1 - 7 1825 9057055
          KF175222.1 - 7 1137
          Gene Symbol: NPIPB9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001287250.1 NP_001274179.1 7 1175
          NM_001287251.1 NP_001274180.1 8 1261
          XM_003118719.7 XP_003118767.2 8 1736
          XM_011545690.2 XP_011543992.1 10 1897
          XM_011545693.2 XP_011543995.1 9 1824
          XM_017022814.1 XP_016878303.1 9 1972
          XM_017022815.1 XP_016878304.1 9 1492
          XM_017022816.1 XP_016878305.1 10 1909
          XM_017022817.1 XP_016878306.1 8 1884
          XM_017022818.1 XP_016878307.1 8 3209
          XM_017022819.1 XP_016878308.1 9 1899
          XM_017022820.1 XP_016878309.1 9 1616
          XM_017022821.1 XP_016878310.1 8 2482
          XM_017022822.1 XP_016878311.1 9 3485
          XM_017022823.1 XP_016878312.1 7 3121
          XM_017022824.1 XP_016878313.1 9 1751
          XM_017022825.1 XP_016878314.1 9 1469
          XM_017022826.1 XP_016878315.1 8 1433
          XM_017022827.1 XP_016878316.1 4 1180
          GenBank mRNA BC171825.1 - 8 1911
          Gene Symbol: PDXDC2P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BX647358.1 - 2 5738

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0