Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDE4DIP SIRNA Assays ›
          Gene Symbol:
          PDE4DIP LOC100996724
          Gene Name:
          phosphodiesterase 4D interacting protein
          phosphodiesterase 4D interacting protein-like
          Gene Aliases:
          CMYA2, MMGL
          -
          Chromosome Location:
          Chr.1: 148808167 - 149051277 on Build GRCh38
          Chromosome Location:
          Chr.1: 120471449 - 120701470 on Build GRCh38
          UniGene ID:
          Hs.584841
          Hs.731111
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s57312

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PDE4DIP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198834.3 NP_001185763.3 24 3694
          NM_014644.5 NP_055459.5 24 3694
          XM_005272981.3 XP_005273038.2 20 4321
          XM_005277442.4 XP_005277499.2 20 4321
          XM_005277443.3 XP_005277500.1 20 4321
          XM_006711642.2 XP_006711705.1 20 4321
          XM_006711643.2 XP_006711706.1 20 4321
          XM_006711644.2 XP_006711707.1 20 4321
          XM_006711646.1 XP_006711709.1 27 3983
          XM_006711650.3 XP_006711713.1 25 3468
          XM_006711651.1 XP_006711714.1 24 3586
          XM_006711652.1 XP_006711715.1 24 3586
          XM_011510172.1 XP_011508474.1 27 4141
          XM_011510173.1 XP_011508475.1 27 4141
          XM_011510174.1 XP_011508476.1 20 4321
          XM_011510175.1 XP_011508477.1 27 4141
          XM_011510179.1 XP_011508481.1 24 3586
          XM_017002875.1 XP_016858364.1 20 4321
          XM_017002876.1 XP_016858365.1 20 4321
          XM_017002877.1 XP_016858366.1 20 4321
          XM_017002878.1 XP_016858367.1 27 4141
          XM_017002879.1 XP_016858368.1 20 4321
          XM_017002880.1 XP_016858369.1 27 4141
          XM_017002881.1 XP_016858370.1 20 4321
          XM_017002882.1 XP_016858371.1 20 4321
          XM_017002883.1 XP_016858372.1 20 4321
          XM_017002884.1 XP_016858373.1 20 4321
          XM_017002887.1 XP_016858376.1 28 4418
          XM_017002888.1 XP_016858377.1 28 4391
          XM_017002889.1 XP_016858378.1 28 4063
          XM_017002890.1 XP_016858379.1 20 4321
          XM_017002891.1 XP_016858380.1 26 4222
          XM_017002892.1 XP_016858381.1 26 3853
          XM_017002893.1 XP_016858382.1 27 4341
          XM_017002894.1 XP_016858383.1 27 3986
          XM_017002895.1 XP_016858384.1 24 3304
          XM_017002896.1 XP_016858385.1 20 4321
          XM_017002897.1 XP_016858386.1 20 4321
          XM_017002898.1 XP_016858387.1 27 3983
          GenBank mRNA AL833273.1 - 24 3582
          BX647660.1 - 24 3622
          Gene Symbol: LOC100996724
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA CR749273.1 - 5 826

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein complex assembly
          protein binding
          enzyme binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline