Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205313.1 | NP_001192242.1 | 10 | 1202 | ||
NM_001205314.1 | NP_001192243.1 | 10 | 1238 | |||
NM_009283.4 | NP_033309.3 | 10 | 1253 | |||
GenBank mRNA | AK018544.1 | - | 10 | 1204 | ||
AK039458.1 | - | 10 | 1254 | |||
AK046517.1 | - | 10 | 1202 | |||
AK082706.1 | - | 10 | 1235 | |||
AK084855.1 | - | 10 | 1187 | |||
AK157263.1 | - | 10 | 1102 | |||
AK159887.1 | - | 10 | 1237 | |||
BC004808.1 | - | 10 | 1189 | 3587831 | ||
U06924.1 | - | 8 | 907 |