Search
Search

Cks1b| Gene Symbol: Cks1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016904.1 | NP_058600.1 | 2 | 194 | ||
| GenBank mRNA | AB025409.1 | - | 2 | 194 | ||
| AK004101.1 | - | 2 | 180 | |||
| AK010414.2 | - | 2 | 181 | |||
| AK011314.1 | - | 2 | 183 | |||
| AK011782.1 | - | 2 | 160 | |||
| BC076579.1 | - | 2 | 186 | 6846517 | ||