Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ammecr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019496.3 | NP_062369.1 | 2 | 548 | ||
GenBank mRNA | AJ243485.1 | - | 2 | 548 | ||
AK051736.1 | - | 2 | 548 |