FIGURE: 1 / 1
The human IL-6 Standard is calibrated against NIBSC IL-6 Reference Standard lot (8)8/514. 1 Thermo pg = 1 NIBSC pg = 0.104 NIBSC units.
Lyophilized product is shipped on cold packs. Stable until expiration if stored desiccated at -20°C. Reconstituted cytokine standards should be stored in aliquots at -70°C. Each aliquot should be used only once and discarded afterwards. Stability of reconstituted and frozen cytokine standards may vary. Controls should be run with each standard curve.
Sufficient For: ELISA standard curve with replicates or as a control in Western blots.
1000 pg/ml when reconstituted at the recommend volume.
Reconstitute with distilled water at volume indicated on label. Use or freeze product within 1 hour of reconstitution.
IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.