Search
This kit permits quick and easy determination of cell viability using two common microscope filters (FITC and RFP) based on intracellular esterase activity and plasma membrane integrity.
1. Culture cells in appropriate medium and vessel for microscopy
2. Thaw vials
3. Add 5 µL calcein AM (Component A) and 20 µL ethidium homodimer-1 (Component B) to 10 mL DPBS to create staining solution.
4. Remove medium from cells
5. Add 100–200 µL of the staining solution directly to cells
6. Incubate 30 minutes at 20–25°C
7. Image cells
| Calcein AM | Ethidium homodimer-1 | |
|---|---|---|
| Excitation/emission | 494/517 nm | 528/617 nm |
| Standard filter set | FITC or GFP | RFP |
| EVOS Light Cube | GFP | RFP |
| Storage conditions | –20°C | –20°C |
Kangaroo rat (PtK2) cells stained with the LIVE/DEAD Viability/Cytotoxicity Kit.