DPBS, calcium, magnesium
Gibco™

DPBS, calcium, magnesium

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityOsmolality
140401171000 mL270 to 300 mOsm/kg
1404018210 x 500 mL270 to 300 mOsm/kg
14040141100 mL270 to 300 mOsm/kg
14040133500 mL270 to 300 mOsm/kg
140402166 x 1000 mL270 to 300 mOsm/kg
Catalog number 14040117
Price (USD)
51.65
Online exclusive
55.00
Save 3.35
Each
Quantity:
1000 mL
Osmolality:
270 to 300 mOsm/kg
Customize this product
Price (USD)
51.65
Online exclusive
55.00
Save 3.35
Each

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:

With: Calcium, Magnesium

Without: Phenol Red

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialDPBS
IncludesNo Sodium Pyruvate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality270 to 300 mOsm/kg
Recommended StorageStorage Conditions: 2°C to 8°C, Shelf Life: 36 months from date of manufacture
SterilitySterile
Sterilization MethodSterile-filtered
Concentration1X
For Use With (Application)Washing and Staining Cells
GradeCell Culture
Physical FormLiquid
Product LineGibco
Quantity1000 mL
Solution TypeCleaning, Storage, pH Buffer
pH7.0 to 7.2
Unit SizeEach

Frequently asked questions (FAQs)

Can I get a specific value for the conductivity for your DPBS products?

Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your DPBS products DNase/RNase-free? Can I use them for RNA work?

Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
An evaluation of a four-channel multiplexed electrospray tandem mass spectrometry for higher throughput quantitative analysis.
Authors: Morrison Denise; Davies Amy E; Watt Alan P;
Journal:Anal Chem
PubMed ID:11985324
A four-channel multiplexed electrospray inlet system (MUX) coupled to a triple quadrupole mass spectrometer was investigated as a higher throughput approach to quantitative analysis. Four discrete samples may be simultaneously analyzed by virtue of a rotating sampler with a concomitant 4-fold increase in analytical throughput. Although absolute sensitivity was reduced ... More