6 Jun 2019
Polymer Beads
Segmentation of the particles located in the foreground of the SEM image.
front
The recipe segments the particles located in the foreground of the image. The same recipe works for beads, powder,... data acquired at different resolution.
A set of markers is defined and then used for a marker-based watershed segmentation. Particles located in the background or touching the boundary of the image are then removed.