26 Jun 2019
Particle Agglomeration
The recipe segments particles that are touching each other, and individual ones.
The recipe segments particles that are touching each other, and individual ones. The recipe creates three main outputs: the individual particles labeled independantly, the agglomerate labeled independantly, and the 2 classified phases (individual and agglomerate).
The area of agglomerate vs individual particles is measured.
The recipe relies on the Filter by Measure Range step that allows to separate agglomerates from single particles based on a roundness measure.