3 Sep 2026
Auto Crop Banner
This module automatically crops the info banner away of the input 2D grayscale image.
Xtra_AutoCropBanner
This module automatically crops the info banner away of the input 2D grayscale image.
It determines the info banner height by scanning the bottom area of the input image.
This search area is set by the parameter 'Height for Sampling' (default value worked for all tested images).
The module generates a duplicate of the input 2D grayscale image but without its info banner that was cropped away.
Limitation: It can only process one 2D grayscale image at a time.
Therefore to process a full stack of 2D images this module must run through a recipe played by Xtra BatchApplyRecipePlayer.