Depthwise Volume Fraction
This module allows the user to compute the volume fraction per separated region depending on depth from the surface of a segmented object.
This module computes the volume fraction per
separated region depending on depth from the surface of a segmented object.
This can be used for trend analysis between samples in the case of depth-wise
structure is important to evaluate such as tablet of medicine or pesticide.
Connections:
Materials [required]
2D/3D label image (Uniform Label Field). Input label volume for
computation of depth-wise volume fraction
Single Object Mask [required]
2D/3D binary label image (Uniform Label Field with 2 labels). Input mask volume
to specify the region for computation
Ports:
Space [unit] [required]
Height of each separated region
Outputs:
A table object stores computed volume fraction per separated regions, distance
map, and labels representing separated regions.