Convert To Spreadsheet
This module converts a field mapped onto an Unstructured Model grid, resulting from a Digital Volume Correlation workflow, into a spreadsheet containing the X,Y,Z positions and the values of the field.
Description:
This module converts field mapped onto an Unstructured Model grid, resulting from a Digital Volume Correlation workflow, into a spreadsheet. The Unstructured Model field mapped onto the grid can be scalar values, 3D vectors or tensors. The resulting spreadsheet will be generated according to the grid's data: it will contain the X,Y,Z positions and the values of the field, eg. displacement, strain, etc. (scalar value for a disp/strain component, Ux,Uy,Uz for a displacement vector, and 6 strain components for a strain tensor field).
Connections:
Data [required]
An Unstructured Model field must be attached. It can be scalar, vector or tensor.
A version of the Xtra compatible with Amira-Avizo Software 2019.3 to 2020.2 can be downloaded HERE.