17 Oct 2019
Eagleford Shale FIB-SEM Segmentation
This recipe guided workflow will result in a 5-phase segmentation applicable to the included dataset and most typical FIB-SEM data. Input Data, the Recipe, Output data are included here.
The Expected input data for this recipe is a 3D FIB-SEM Stack. This recipe is optimized for rock data especially carbonate rich samples such as chalks, marls and marly shales. The resolution of the data is in itself not important so long as the porosity and other phases of interest are fully resolved.