24 Aug 2020
Selective Border Kill
A module that allows you to filter particles selectively, based on their contact with each border of the image.
This module is similar to the Border Kill module, but adds options that allow you to filter out (or keep) objects that either touch or do not touch specific borders of the image.