8 Jul 2026
How to Visualize 3D Image Data
Amira and Avizo provide several ways to view 3D image data. This video explains how to view 3D image data as a series of 2D Ortho Slices, as an Isosurface, and as a direct 3D Volume Rendering.
This tutorial shows how to view 3D images in a 3D viewer within Amira and Avizo Software. It covers how to use Volume Rendering, create Isosurfaces, and how to manage multiple Ortho Slices. An introduction to colormaps is provided.